3-Phenylaniline ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Aminobiphenyl, 3-Biphenylylamine, [1,1'-Biphenyl]-3-amine, Biphenyl-3-ylamine

B0987-5G B0987-1G B0987-25G
TCB0987-005GEA 230.68 CAD
TCB0987-005G TCB0987-001G TCB0987-25G
3-Phenylaniline ≥99.0% (by GC, titration analysis)
3-Phenylaniline

CAS Number: 2243-47-2
MDL Number: MFCD00047846
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 254
Flash Point (°C): 110
Freezing point (°C): 29


Other Product Names / Synonyms: 3-Aminodiphenyl, 3-Biphenylamine

Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 254 °C
Melting Pt: 33 °C
Flash Pt: 110 °C
MDL Number: MFCD00047846
CAS Number: 2243-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ed7b7a1f7a9b6362c8dde38f4ae7678442d0d49&vid=8ed7b7a1f7a9b6362c8dde38f4ae7678442d0d49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR