5-Amino-1-naphthol-3-sulfonic Acid Hydrate ≥97.0%
Supplier: TCI America
Synonyms:
8-Amino-4-hydroxy-2-naphthalenesulphonic Acid, M Acid, 8-Amino-4-hydroxy-2-naphthalenesulfonic Acid
A0368-25G
A0368-500G
TCA0368-025GEA
50.56
CAD
TCA0368-025G
TCA0368-500G
5-Amino-1-naphthol-3-sulfonic Acid Hydrate ≥97.0%
5-Amino-1-naphthol-3-sulfonic Acid Hydrate
CAS Number: 489-78-1
MDL Number: MFCD00035712
Molecular Formula: C10H9NO4S
Molecular Weight: 239.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Other Product Names / Synonyms: 8-Amino-4-hydroxy-2-naphthalenesulfonic Acid, M Acid
Formula:
C₁₀H₉NO₄S MW: 239.25 g/mol |
MDL Number:
MFCD00035712 CAS Number: 489-78-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...