5-Hydroxy-4-octanone ≥96.0% for synthesis

Supplier: TCI America

Synonyms: 5-Hydroxyoctan-4-one, 5-Hydroxy-4-octanone

B0971-25ML B0971-5ML
TCB0971-025MLEA 297.04 CAD
TCB0971-025ML TCB0971-5ML
5-Hydroxy-4-octanone ≥96.0% for synthesis
5-Hydroxy-4-octanone

CAS Number: 496-77-5
MDL Number: MFCD00021928
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 77
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 5-Hydroxy-4-octanone

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 188
Melting Pt: 2
Density: 0.92 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00021928
CAS Number: 496-77-5
EINECS: 207-830-4
Merck Index: 13,01595

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR