2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres) ≥93.0%
Supplier: TCI America
|
Warning
|
B0928-25G
B0928-500G
TCB0928-025GEA
42.66
CAD
TCB0928-025G
TCB0928-500G
2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres) ≥93.0%
2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres)
CAS Number: 80-04-6
MDL Number: MFCD00019334
Molecular Formula: C15H28O2
Molecular Weight: 240.39
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Flash Point (°C): 110
Other Product Names / Synonyms: p,p'-Dihydroxy-2,2-dicyclohexylpropane, Perhydrobisphenol A
Formula:
C₁₅H₂₈O₂ Flash Pt: 110 °C |
MDL Number:
MFCD00019334 CAS Number: 80-04-6 EINECS: 201-244-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...