2-Amino-1-naphthol hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
A0365-1G
A0365-5G
TCA0365-001GEA
143.78
CAD
TCA0365-001G
TCA0365-005G
2-Amino-1-naphthol hydrochloride ≥98.0% (by total nitrogen and titration analysis)
2-Amino-1-naphthol hydrochloride
CAS Number: 41772-23-0
MDL Number: MFCD00050530
Molecular Formula: C10H9NO
Molecular Weight: 195.65
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Formula:
C₁₀H₁₀ClNO |
MDL Number:
MFCD00050530 CAS Number: 41772-23-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...