1,1'-Dibenzoylferrocene ≥98.0%
Supplier: TCI America
B0915-5G
B0915-25G
TCB0915-005GEA
107.44
CAD
TCB0915-005G
TCB0915-025G
1,1'-Dibenzoylferrocene ≥98.0%
1,1'-Dibenzoylferrocene
CAS Number: 12180-80-2
MDL Number: MFCD00045455
Molecular Formula: C24H18FeO2
Molecular Weight: 394.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103
Other Product Names / Synonyms: Bis(benzoylcyclopentadienyl) Iron, Di(benzoylcyclopentadienyl) Iron
Formula:
C₂₄H₁₀FeO₂ Melting Pt: 103 °C |
MDL Number:
MFCD00045455 CAS Number: 12180-80-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...