1-Amino-8-naphthol-2,4-disulfonic acid monosodium salt hydrate ≥85.0%
Supplier: TCI America
Synonyms:
SS acid, Monosodium 1-Amino-8-naphthol-2,4-disulfonate, Monosodium 1-Amino-8-naphthol-2,4-disulphonate, 2S acid, Chicago acid, 4-Amino-5-hydroxy-1,3-naphthalenedisulfonic acid Monosodium salt, Sodium Hydrogen 4-Amino-5-hydroxy-1,3-naphthalenedisulfonate, 4-Amino-5-hydroxy-1,3-naphthalenedisulphonic acid Monosodium salt, Sodium Hydrogen 4-Amino-5-hydroxy-1,3-naphthalenedisulphonate
CAS Number: 52789-62-5
MDL Number: MFCD00068275
Molecular Formula: C10H9NO7S2
Molecular Weight: 341.28
Purity/Analysis Method: >85.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 4-Amino-5-hydroxy-1,3-naphthalenedisulfonic Acid Monosodium Salt, Chicago Acid, 2S Acid, Sodium Hydrogen 4-Amino-5-hydroxy-1,3-naphthalenedisulfonate, SS Acid, Monosodium 1-Amino-8-naphthol-2,4-disulfonate
Formula:
C₁₀H₉NO₇S₂ MW: 341.28 g/mol |
MDL Number:
MFCD00068275 CAS Number: 52789-62-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...