Terephthalbis(p-phenetidine) ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: Di-p-phenetidylterephthalaldehyde, Bisphenetidylterephthalaldehyde, Bis(p-ethoxyanilino)terephthalaldehyde

B0871-25G B0871-5G
TCB0871-25GEA 395 CAD
TCB0871-25G TCB0871-5G
Terephthalbis(p-phenetidine) ≥98.0% (by GC, total nitrogen)
Terephthalbis(p-phenetidine)

CAS Number: 17696-60-5
MDL Number: MFCD00026796
Molecular Formula: C24H24N2O2
Molecular Weight: 372.47
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: Bis(p-ethoxyanilino)terephthalaldehyde, Bisphenetidylterephthalaldehyde, Di-p-phenetidylterephthalaldehyde

Formula: C₂₄H₂₄N₂O₂
MW: 372.47 g/mol
MDL Number: MFCD00026796
CAS Number: 17696-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=182ce2ad1be8a34622489f3232b8642defb3f827&vid=182ce2ad1be8a34622489f3232b8642defb3f827&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR