4-Bromo-1,8-naphthalic anhydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

B0858-25G B0858-5G
TCB0858-025GEA 287.24 CAD
TCB0858-025G TCB0858-005G
4-Bromo-1,8-naphthalic anhydride ≥95.0% (by titrimetric analysis)
4-Bromo-1,8-naphthalic anhydride

CAS Number: 81-86-7
MDL Number: MFCD00006927
Molecular Formula: C12H5BrO3
Molecular Weight: 277.07
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 221


Other Product Names / Synonyms: 4-Bromo-1,8-naphthalenedicarboxylic Anhydride

Formula: C₁₂H₅BrO₃
Melting Pt: 221 °C
MDL Number: MFCD00006927
CAS Number: 81-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99436a82745d1925b86328bc604b93c6295568c&vid=f99436a82745d1925b86328bc604b93c6295568c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR