Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B0853-25G
B0853-5G
TCB0853-025GEA
262.28
CAD
TCB0853-025G
TCB0853-005G
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride
CAS Number: 2645-08-1
MDL Number: MFCD00012579
Molecular Formula: C15H22N4O3
Molecular Weight: 342.82
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 130
Specific rotation [a]20/D: -17 deg (C=2, H2O)
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Bz-Arg-OEt.HCl
Formula:
C₁₅H₂₂N₄O₃ Melting Pt: 130 °C |
MDL Number:
MFCD00012579 CAS Number: 2645-08-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...