6-Amino-1-naphthol ≥98.0% (by HPLC)

Supplier: TCI America

A0360-5G A0360-1G
TCA0360-005GEA 630.42 CAD
TCA0360-005G TCA0360-001G
6-Amino-1-naphthol ≥98.0% (by HPLC)
6-Amino-1-naphthol

CAS Number: 23894-12-4
MDL Number: MFCD00059543
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 206

Formula: C₁₀H₉NO
Melting Pt: 206 °C
MDL Number: MFCD00059543
CAS Number: 23894-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e091c531e2a90af598bf2424f936b8fcd0458cd4&vid=e091c531e2a90af598bf2424f936b8fcd0458cd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR