6,6'-Dimethyl-2,2'-bipyridyl ≥98.0%

Supplier: TCI America

Synonyms: 6,6'-Dimethyl-2,2'-bipyridine

B0842-1G B0842-5G B0842-100MG
TCB0842-1GEA 132.72 CAD
TCB0842-1G TCB0842-5G TCB0842-100MG
6,6'-Dimethyl-2,2'-bipyridyl ≥98.0%
6,6'-Dimethyl-2,2'-bipyridyl

CAS Number: 4411-80-7
MDL Number: MFCD00059776
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 286
Melting point (°C): 88


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 6,6'-Bi-2-picoline, 6,6'-Di-2-picolyl

Formula: C₁₂H₁₂N₂
MW: 184.2371 g/mol
Boiling Pt: 286 °C
Melting Pt: 88 °C
MDL Number: MFCD00059776
CAS Number: 4411-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba3e1a54a746fe9889684e324bd11bea59c2fa12&vid=ba3e1a54a746fe9889684e324bd11bea59c2fa12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR