Diethyl-4,4'-biphenyldicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Biphenyldicarboxylic Acid Diethyl Ester, 4,4'-Bibenzoic Acid Diethyl Ester, Diethyl 4,4'-Bibenzoate
CAS Number: 47230-38-6
MDL Number: MFCD00017270
Molecular Formula: C18H18O4
Molecular Weight: 298.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113
Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Biphenyldicarboxylic Acid Diethyl Ester, Diethyl 4,4'-Bibenzoate, 4,4'-Bibenzoic Acid Diethyl Ester
Formula:
C₁₈H₁₈O₄ Melting Pt: 113 °C |
MDL Number:
MFCD00017270 CAS Number: 47230-38-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...