Bis(4-chlorophenyl) sulfone ≥98.0%

Supplier: TCI America

B0810-500G B0810-25G B0810-100G
TCB0810-500GEA 149.94 CAD
TCB0810-500G TCB0810-025G TCB0810-100G
Bis(4-chlorophenyl) sulfone ≥98.0%
Bis(4-chlorophenyl) sulphone

CAS Number: 80-07-9
MDL Number: MFCD00000619
Molecular Formula: C12H8Cl2O2S
Molecular Weight: 287.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 148


Product Categories: Materials Chemistry, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(4-chlorophenyl) Sulfone

Formula: C₁₂H₈Cl₂O₂S
Melting Pt: 148 °C
MDL Number: MFCD00000619
CAS Number: 80-07-9
EINECS: 201-247-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e89d1aaeb7afff5681e9669a6a41e710d552edd4&vid=e89d1aaeb7afff5681e9669a6a41e710d552edd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR