(±)-1,2-Butanediol ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Dihydroxybutane, (±)-1,2-Butylene glycol, (±)-1,2-Dihydroxy butane, 1,2-Butylene glycol, Butane-1,2-diol, 1,2-Butanediol

B0804-25G B0804-500G
TCB0804-025GEA 33.76 CAD
TCB0804-025G TCB0804-500G
(±)-1,2-Butanediol ≥98.0%
(±)-1,2-Butanediol

CAS Number: 584-03-2
MDL Number: MFCD00004570
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 110
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 1,2-Butylene Glycol, 1,2-Dihydroxybutane

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 194 °C
Melting Pt: –50 °C
Density: 1.00 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004570
CAS Number: 584-03-2
EINECS: 209-527-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dff144a2140af5c179af439cbf9b0908458d42b&vid=7dff144a2140af5c179af439cbf9b0908458d42b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR