Sodium-7-amino-2-naphthalenesulfonate ≥70.0%
Supplier: TCI America
Synonyms:
Cassella's acid F Sodium Salt, 7-Amino-2-naphthalenesulphonic acid Sodium Salt, 2-Naphthylamine-7-sulphonic acid Sodium Salt, 2-Naphthylamine-7-sulfonic acid Sodium Salt, 7-Amino-2-naphthalenesulfonic acid Sodium Salt
CAS Number: 5412-82-8
MDL Number: MFCD00053436
Molecular Formula: C10H9NO3S
Molecular Weight: 245.23
Purity/Analysis Method: >70.0% (N)
Form: Crystal
Other Product Names / Synonyms: Cassella's Acid F Sodium Salt, 2-Naphthylamine-7-sulfonic Acid Sodium Salt, 7-Amino-2-naphthalenesulfonic Acid Sodium Salt
Formula:
C₁₀H₉NO₃S MW: 245.23 g/mol |
MDL Number:
MFCD00053436 CAS Number: 5412-82-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...