Butyl phthalyl butyl glycolate ≥95.0%

Supplier: TCI America

Synonyms: Phthalic Acid Butoxycarbonylmethyl Butyl Ester, Butoxycarbonylmethyl Butyl Phthalate

B0737-500G B0737-25G
TCB0737-500GEA 274.45 CAD
TCB0737-500G TCB0737-025G
Butyl phthalyl butyl glycolate ≥95.0%
Butyl phthalyl butyl glycolate

CAS Number: 85-70-1
MDL Number: MFCD00053793
Molecular Formula: C18H24O6
Molecular Weight: 336.38
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 345
Flash Point (°C): 199
Specific Gravity (20/20): 1.10


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Butoxycarbonylmethyl Butyl Phthalate, Phthalic Acid Butoxycarbonylmethyl Butyl Ester

Formula: C₁₈H₂₄O₆
MW: 336.38 g/mol
Boiling Pt: 345 °C
Density: 1.10 (20 °C)
Flash Pt: 199 °C
MDL Number: MFCD00053793
CAS Number: 85-70-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05c5a258ab042df45393ce6c434277d2a89defa2&vid=05c5a258ab042df45393ce6c434277d2a89defa2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR