2,4-Diamino-6-butylamino-1,3,5-triazine ≥97.0%

Supplier: TCI America

Synonyms: N2-Butyl-1,3,5-triazine-2,4,6-triamine, N2-Butylmelamine, 2-Butylamino-4,6-diamino-1,3,5-triazine

B0727-5G B0727-25G
TCB0727-005GEA 75.84 CAD
TCB0727-005G TCB0727-025G
2,4-Diamino-6-butylamino-1,3,5-triazine ≥97.0%
2,4-Diamino-6-butylamino-1,3,5-triazine

CAS Number: 5606-24-6
MDL Number: MFCD00059764
Molecular Formula: C7H14N6
Molecular Weight: 182.23
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Butylamino-4,6-diamino-1,3,5-triazine, N2-Butylmelamine

Formula: C₇H₁₄N₆
MW: 182.23 g/mol
MDL Number: MFCD00059764
CAS Number: 5606-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e43ee86aa8f986de0742de84c61e8a329e0b2d67&vid=e43ee86aa8f986de0742de84c61e8a329e0b2d67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR