4,4'-Butylidenebis(6-tert-butyl-m-cresol) ≥97.0%

Supplier: TCI America

Synonyms: 4,4'-(Butane-1,1-diyl)bis(2-(tert-butyl)-5-methylphenol)

B0724-500G B0724-25G
TCB0724-500GEA 366.56 CAD
TCB0724-500G TCB0724-025G
4,4'-Butylidenebis(6-tert-butyl-m-cresol) ≥97.0%
4,4'-Butylidenebis(6-tert-butyl-m-cresol)

CAS Number: 85-60-9
MDL Number: MFCD00026289
Molecular Formula: C26H38O2
Molecular Weight: 382.59
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 211

Formula: C₂₆H₃₈O₂
Melting Pt: 211 °C
MDL Number: MFCD00026289
CAS Number: 85-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9814edef4390ebb8b4d26ec5070253de68395237&vid=9814edef4390ebb8b4d26ec5070253de68395237&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR