Butylamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Butan-1-amine hydrochloride, 1-Aminobutane Hydrochloride
B0710-25G
B0710-500G
TCB0710-025GEA
66.36
CAD
TCB0710-025G
TCB0710-500G
Butylamine hydrochloride ≥98.0% (by titrimetric analysis)
Butylamine hydrochloride
CAS Number: 3858-78-4
MDL Number: MFCD00043791
Molecular Formula: C4H11N
Molecular Weight: 109.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 214
Other Product Names / Synonyms: 1-Aminobutane Hydrochloride
Formula:
C₄H₁₁N MW: 109.60 g/mol Melting Pt: 214 °C |
MDL Number:
MFCD00043791 CAS Number: 3858-78-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...