3-Butoxyphenol ≥96.0%

Supplier: TCI America

Synonyms: Resorcinol Monobutyl Ether

B0701-25G B0701-5G
TCB0701-025GEA 454.57 CAD
TCB0701-025G TCB0701-5G
3-Butoxyphenol ≥96.0%
3-Butoxyphenol

CAS Number: 18979-72-1
MDL Number: MFCD00058976
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 269
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Resorcinol Monobutyl Ether

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 269 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00058976
CAS Number: 18979-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=688365bf1acfcd0d97dbe9c1a3f813c94890ddcb&vid=688365bf1acfcd0d97dbe9c1a3f813c94890ddcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR