6-Amino-2-naphthalenesulfonic acid monohydrate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Bronner's acid, 2-Naphthylamine-6-sulfonic acid, 2-Naphthylamine-6-sulphonic acid
A0348-25G
A0348-500G
TCA0348-025GEA
96.38
CAD
TCA0348-025G
TCA0348-500G
6-Amino-2-naphthalenesulfonic acid monohydrate ≥98.0% (by HPLC)
6-Amino-2-naphthalenesulfonic acid monohydrate
CAS Number: 93-00-5
MDL Number: MFCD04038015
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Other Product Names / Synonyms: Bronner's Acid, 2-Naphthylamine-6-sulfonic Acid
Formula:
C₁₀H₉NO₃S MW: 223.25 g/mol |
MDL Number:
MFCD04038015 CAS Number: 93-00-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...