3'-Methylacetanilide ≥98.0%

Supplier: TCI America

Synonyms: m-Acetotoluidide, m-Acetotoluidid, 3-Methylacetanilide, N-(3-Methylphenyl)acetamide

A0062-25G A0062-250G
TCA0062-025GEA 96.38 CAD
TCA0062-025G TCA0062-250G
3'-Methylacetanilide ≥98.0%
3'-Methylacetanilide

CAS Number: 537-92-8
MDL Number: MFCD00014962
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 163
Melting point (°C): 66


Other Product Names / Synonyms: N-Acetyl-m-toluidine, 3'-Methylacetanilide

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 163 °C
Melting Pt: 66 °C
Density: 1.141 g/cm³ (15 °C)
MDL Number: MFCD00014962
CAS Number: 537-92-8
EINECS: 208-678-1
Merck Index: 13,00076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3a8399eb01c7b7846d36c149883d422832f059e&vid=c3a8399eb01c7b7846d36c149883d422832f059e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR