Xanthan gum
Supplier: TCI America
X0048-25G
X0048-500G
X0048-100G
TCX0048-025GEA
101.06
CAD
TCX0048-025G
TCX0048-500G
TCX0048-100G
Xanthan gum
Xanthan gum
CAS Number: 11138-66-2
MDL Number: MFCD00131256
Form: Crystal
Color: Very Pale Yellow
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides
Density:
1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00131256 CAS Number: 11138-66-2 EINECS: 234-394-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...