Xanthene-9-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 9H-Xanthene-9-carboxylic acid

X0031-5G X0031-25G
TCX0031-005GEA 66.36 CAD
TCX0031-005G TCX0031-025G
Xanthene-9-carboxylic acid ≥98.0%
Xanthene-9-carboxylic acid

CAS Number: 82-07-5
MDL Number: MFCD00005059
Molecular Formula: C14H10O3
Molecular Weight: 226.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 214

Formula: C₁₄H₁₀O₃
Melting Pt: 214 °C
MDL Number: MFCD00005059
CAS Number: 82-07-5
EINECS: 201-394-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b0cf347ebc613c05104a56d8c73b37155e0bf3b&vid=5b0cf347ebc613c05104a56d8c73b37155e0bf3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR