trans-2-Undecenal ≥93.0%

Supplier: TCI America

U0046-5ML U0046-25ML
TCU0046-005MLEA 56.88 CAD
TCU0046-005ML TCU0046-25ML
trans-2-Undecenal ≥93.0%
trans-2-Undecenal

CAS Number: 53448-07-0
MDL Number: MFCD00014680
Molecular Formula: C11H20O
Molecular Weight: 168.28
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 240
Flash Point (°C): 120
Specific Gravity (20/20): 0.84

Formula: C₁₁H₂₀O
Boiling Pt: 240 °C
Density: 0.84 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00014680
CAS Number: 53448-07-0
EINECS: 258-559-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b5156b0c6e7c291ebf1106e421d184eb609f123&vid=7b5156b0c6e7c291ebf1106e421d184eb609f123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR