6-Undecanone ≥98.0%

Supplier: TCI America

U0043-25ML U0043-100ML
TCU0043-025MLEA 115.34 CAD
TCU0043-025ML TCU0043-100ML
6-Undecanone ≥98.0%
6-Undecanone

CAS Number: 927-49-1
MDL Number: MFCD00009516
Molecular Formula: C11H22O
Molecular Weight: 170.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 226
Flash Point (°C): 88
Freezing point (°C): 15
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Diamyl Ketone, Dipentyl Ketone

Formula: C₁₁H₂₂O
Boiling Pt: 226 °C
Melting Pt: 15.5 °C
Density: 0.83 (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00009516
CAS Number: 927-49-1
EINECS: 213-150-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6092e1e93002c629f5797cc5d9daa08969d515d6&vid=6092e1e93002c629f5797cc5d9daa08969d515d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR