Methyl-10-undecenoate ≥96.0%

Supplier: TCI America

Synonyms: Undecenylic acid methyl ester, 10-Undecenoic acid methyl ester, Methyl undec-10-enoate, Undec-10-enoic acid emthyl ester

U0036-25ML U0036-500ML U0036-100ML
TCU0036-025MLEA 60.12 CAD
TCU0036-025ML TCU0036-500ML TCU0036-100ML
Methyl-10-undecenoate ≥96.0%
Methyl 10-undecenoate

CAS Number: 111-81-9
MDL Number: MFCD00016689
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 248
Flash Point (°C): 122
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: 10-Undecenoic Acid Methyl Ester

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Boiling Pt: 248 °C
Melting Pt: –27.5 °C
Density: 0.89 (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00016689
CAS Number: 111-81-9
EINECS: 203-910-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d674a28b1403e1e3afa4de79720960d30414d094&vid=d674a28b1403e1e3afa4de79720960d30414d094&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR