Uridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

U0020-5G U0020-25G
TCU0020-005GEA 47.13 CAD
TCU0020-005G TCU0020-25G
Uridine ≥98.0% (by titrimetric analysis)
Uridine

CAS Number: 58-96-8
MDL Number: MFCD00006526
Molecular Formula: C9H12N2O6
Molecular Weight: 244.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 165
Specific rotation [a]20/D: 9 deg (C=2, H2O)

Formula: C₉H₁₂N₂O₆
Melting Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS Number: 58-96-8
EINECS: 200-407-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a324c3416862b10a8557346e3935bd0e11b0e4f9&vid=a324c3416862b10a8557346e3935bd0e11b0e4f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR