2,4,5-Trimethylbenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T2469-25G T2469-5G
TCT2469-25GEA 785.26 CAD
TCT2469-25G TCT2469-5G
2,4,5-Trimethylbenzoic acid ≥97.0% (by GC, titration analysis)
2,4,5-Trimethylbenzoic acid

CAS Number: 528-90-5
MDL Number: MFCD00075863
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 151

Formula: C₁₀H₁₂O₂
Melting Pt: 151 °C
MDL Number: MFCD00075863
CAS Number: 528-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cc03026d9062f55c72dac3af2dbbd79805f2d6e&vid=4cc03026d9062f55c72dac3af2dbbd79805f2d6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR