(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol ≥98.0%
Supplier: TCI America
Synonyms:
(S)-1,2,3,4-Tetrahydronaphthalen-1-ol, (S)-(+)-alpha-Tetralol
CAS Number: 53732-47-1
MDL Number: MFCD00063006
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 92
Melting point (°C): 41
Flash Point (°C): 113
Specific rotation [a]20/D: 33 deg (C=2.5, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-alpha-Tetralol
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 92 °C Melting Pt: 41 °C Flash Pt: 113 °C |
MDL Number:
MFCD00063006 CAS Number: 53732-47-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...