(R)-(-)-1,2,3,4-Tetrahydro-1-naphthol ≥98.0%
Supplier: TCI America
Synonyms:
(R)-1,2,3,4-Tetrahydronaphthalen-1-ol, (R)-(-)-alpha-Tetralol
CAS Number: 23357-45-1
MDL Number: MFCD00063005
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 140
Melting point (°C): 40
Flash Point (°C): 113
Specific rotation [a]20/D: -32 deg (C=2.5, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-alpha-Tetralol
Formula:
C₁₀H₁₂O Boiling Pt: 140 °C Melting Pt: 40 °C Flash Pt: 113 °C |
MDL Number:
MFCD00063005 CAS Number: 23357-45-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...