2,4,6-Tribromoanisole ≥98.0%

Supplier: TCI America

T2301-25G T2301-5G
TCT2301-25GEA 276.03 CAD
TCT2301-25G TCT2301-5G
2,4,6-Tribromoanisole ≥98.0%
2,4,6-Tribromoanisole

CAS Number: 607-99-8
MDL Number: MFCD00192510
Molecular Formula: C7H5Br3O
Molecular Weight: 344.83
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 299
Melting point (°C): 88

Formula: C₇H₅Br₃O
Boiling Pt: 299 °C
Melting Pt: 88 °C
Density: 2.13 g/cm³ (20 °C)
MDL Number: MFCD00192510
CAS Number: 607-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8a8932bc0cd282d451b591211354630d50a2277&vid=c8a8932bc0cd282d451b591211354630d50a2277&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR