2,4,6-Trimethoxypyrimidine ≥98.0%
Supplier: TCI America
T2039-1G
T2039-5G
TCT2039-1GEA
75.84
CAD
TCT2039-1G
TCT2039-5G
2,4,6-Trimethoxypyrimidine ≥98.0%
2,4,6-Trimethoxypyrimidine
CAS Number: 13106-85-9
MDL Number: MFCD00829225
Molecular Formula: C7H10N2O3
Molecular Weight: 170.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 232
Melting point (°C): 54
Formula:
C₇H₁₀N₂O₃ Boiling Pt: 232 °C Melting Pt: 54 °C |
MDL Number:
MFCD00829225 CAS Number: 13106-85-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...