2,3,5,6-Tetrafluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

T1881-5G T1881-1G
TCT1881-5GEA 94.8 CAD
TCT1881-5G 76220-512
2,3,5,6-Tetrafluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,5,6-Tetrafluorobenzoic acid

CAS Number: 652-18-6
MDL Number: MFCD00002408
Molecular Formula: C7H2F4O2
Molecular Weight: 194.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 150

Formula: C₇H₂F₄O₂
MW: 194.09 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00002408
CAS Number: 652-18-6
EINECS: 416-800-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4cfc563837cab1cbf78515fe1c2d61c8d22accb&vid=a4cfc563837cab1cbf78515fe1c2d61c8d22accb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR