1H,1H-Tridecafluoro-1-heptanol ≥95.0%

Supplier: TCI America

T1701-5G T1701-25G
TCT1701-005GEA 139.04 CAD
TCT1701-005G TCT1701-25G
1H,1H-Tridecafluoro-1-heptanol ≥95.0%
1H,1H-Tridecafluoro-1-heptanol

CAS Number: 375-82-6
MDL Number: MFCD00042433
Molecular Formula: C7H3F13O
Molecular Weight: 350.08
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 95
Specific Gravity (20/20): 1.75


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Perfluoro-1-heptanol

Formula: C₇H₃F₁₃O
Boiling Pt: 95 °C
Density: 1.75 (20 °C)
MDL Number: MFCD00042433
CAS Number: 375-82-6
EINECS: 206-796-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc66ffbb0e8e68096f434081f55746c4d0a16b3&vid=cbc66ffbb0e8e68096f434081f55746c4d0a16b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR