Tetraethylphosphonium bromide ≥98.0%

Supplier: TCI America

T1379-5G T1379-25G
TCT1379-005GEA 55.62 CAD
TCT1379-005G TCT1379-025G
Tetraethylphosphonium bromide ≥98.0%
Tetraethylphosphonium bromide

CAS Number: 4317-07-1
MDL Number: MFCD00054230
Molecular Formula: C8H20BrP
Molecular Weight: 227.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₈H₂₀BrP
MDL Number: MFCD00054230
CAS Number: 4317-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c0187fc6250a868492db3b6e44aa2f3d39ef7e8&vid=2c0187fc6250a868492db3b6e44aa2f3d39ef7e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR