Ethyl-4,4,4-trichloroacetoacetate ≥97.0%

Supplier: TCI America

T1285-25G T1285-10G
TCT1285-25GEA 634.69 CAD
TCT1285-25G TCT1285-010G
Ethyl-4,4,4-trichloroacetoacetate ≥97.0%
Ethyl 4,4,4-trichloroacetoacetate

CAS Number: 3702-98-5
MDL Number: MFCD00043329
Molecular Formula: C6H7Cl3O3
Molecular Weight: 233.47
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 234
Specific Gravity (20/20): 1.39


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: 4,4,4-Trichloroacetoacetic Acid Ethyl Ester

Formula: C₆H₇Cl₃O₃
Boiling Pt: 234 °C
Density: 1.39 (20 °C)
MDL Number: MFCD00043329
CAS Number: 3702-98-5
EINECS: 223-041-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ca058aee52401379c3a0b935cbb2c6239811443&vid=1ca058aee52401379c3a0b935cbb2c6239811443&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR