Tetradecylcyclohexane ≥98.0%

Supplier: TCI America

T0873-25G T0873-5G
TCT0873-025GEA 1594.22 CAD
TCT0873-025G TCT0873-005G
Tetradecylcyclohexane ≥98.0%
Tetradecylcyclohexane

CAS Number: 1795-18-2
MDL Number: MFCD00058951
Molecular Formula: C20H40
Molecular Weight: 280.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Freezing point (°C): 24


Other Product Names / Synonyms: Myristylcyclohexane, 1-Cyclohexyltetradecane

Formula: C₂₀H₄₀
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 42 °C
Density: 0.8254 g/cm³ (20 °C)
MDL Number: MFCD00058951
CAS Number: 1795-18-2
EINECS: 217-274-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10ef0b957a7d61aace986e1cf3a7f67f2bb7797b&vid=10ef0b957a7d61aace986e1cf3a7f67f2bb7797b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR