2,4,6-Trichloroanisole ≥98.0%

Supplier: TCI America
Warning

T0867-5G T0867-25G
TCT0867-005GEA 140.62 CAD
TCT0867-005G TCT0867-025G
2,4,6-Trichloroanisole ≥98.0%
2,4,6-Trichloroanisole

CAS Number: 87-40-1
MDL Number: MFCD00000588
Molecular Formula: C7H5Cl3O
Molecular Weight: 211.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 240
Melting point (°C): 60


Other Product Names / Synonyms: 2,4,6-Trichloro-1-methoxybenzene

Formula: C₇H₅Cl₃O
Boiling Pt: 240 °C
Melting Pt: 60 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000588
CAS Number: 87-40-1
EINECS: 201-743-5
Merck Index: 13,09702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4103f8e0c91832b58a8aa9c7d0bd51f87fb026dd&vid=507c3409a53c1edbba1ae19201ec7207cd923690&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR