Methyl triacontanate ≥95.0%

Supplier: TCI America

Synonyms: Mellisic acid methyl ester, Triacontanoic acid methyl ester

T0812-1G T0812-100MG
TCT0812-1GEA 1377.76 CAD
TCT0812-1G TCT0812-100MG
Methyl triacontanate ≥95.0%
Methyl triacontanate

CAS Number: 629-83-4
MDL Number: MFCD00042897
Molecular Formula: C31H62O2
Molecular Weight: 466.84
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 72
Flash Point (°C): 110


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Melissic Acid Methyl Ester, Methyl Melissate, Triacontanoic Acid Methyl Ester

Formula: C₃₁H₆₂O₂
Melting Pt: 72 °C
Flash Pt: 110 °C
MDL Number: MFCD00042897
CAS Number: 629-83-4
EINECS: 211-113-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7f5e6dafdc086d35fed3d71681e16e75a47302c&vid=b7f5e6dafdc086d35fed3d71681e16e75a47302c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR