2,3,4-Trichloroaniline ≥98.0%
Supplier: TCI America
T0798-25G
T0798-5G
TCT0798-025GEA
843.72
CAD
TCT0798-025G
TCT0798-5G
2,3,4-Trichloroaniline ≥98.0%
2,3,4-Trichloroaniline
CAS Number: 634-67-3
MDL Number: MFCD00042819
Molecular Formula: C6H4Cl3N
Molecular Weight: 196.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 292
Melting point (°C): 67
Formula:
C₆H₄Cl₃N Boiling Pt: 292 °C Melting Pt: 67 °C Density: 1.54 g/cm³ (20 °C) |
MDL Number:
MFCD00042819 CAS Number: 634-67-3 EINECS: 211-215-6 UN: 2811 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...