Triethylene glycol monomethyl ether ≥98.0%

Supplier: TCI America

T0709-500ML T0709-25ML
TCT0709-500MLEA 97.94 CAD
TCT0709-500ML TCT0709-025ML
Triethylene glycol monomethyl ether ≥98.0%
Triethylene glycol monomethyl ether

CAS Number: 112-35-6
MDL Number: MFCD00002875
Molecular Formula: C7H16O4
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 248
Melting point (°C): -45
Flash Point (°C): 134
Specific Gravity (20/20): 1.05


Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: Methyl Triglycol, mPEG3-Alcohol

Formula: C₇H₁₆O₄
Boiling Pt: 248 °C
Melting Pt: –45 °C
Density: 1.05 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00002875
CAS Number: 112-35-6
EINECS: 203-962-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ebd90d0ff453f551c98836978381f1d0045be4b&vid=0ebd90d0ff453f551c98836978381f1d0045be4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR