1,3-Diaminopropane dihydrochloride ≥97.0%

Supplier: TCI America

Synonyms: Propane-1,3-diamine dihydrochloride, 1,3-Propanediamine dihydrochloride, Trimethylenediamine dihydrochloride

T0613-25G T0613-100G
TCT0613-025GEA 77.42 CAD
TCT0613-025G 76220-490
1,3-Diaminopropane dihydrochloride ≥97.0%
1,3-Diaminopropane dihydrochloride

CAS Number: 10517-44-9
MDL Number: MFCD00012525
Molecular Formula: C3H10N2
Molecular Weight: 147.04
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 253


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Trimethylenediamine Dihydrochloride, 1,3-Propanediamine Dihydrochloride

Formula: C₃H₁₂Cl₂N₂
MW: 147.05 g/mol
Melting Pt: 253 °C
MDL Number: MFCD00012525
CAS Number: 10517-44-9
EINECS: 234-051-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d44ff25251f62c94b6e1698cc72e2cb27e748976&vid=d44ff25251f62c94b6e1698cc72e2cb27e748976&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR