3,3,5-Trimethylcyclohexanone ≥98.0%
Supplier: TCI America
T0602-25ML
T0602-500ML
TCT0602-025MLEA
31.6
CAD
TCT0602-025ML
TCT0602-500ML
3,3,5-Trimethylcyclohexanone ≥98.0%
3,3,5-Trimethylcyclohexanone
CAS Number: 873-94-9
MDL Number: MFCD00019466
Molecular Formula: C9H16O
Molecular Weight: 140.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Flash Point (°C): 68
Specific Gravity (20/20): 0.89
Formula:
C₉H₁₆O MW: 140.23 g/mol Boiling Pt: 190 °C Melting Pt: –10 °C Density: 0.89 (20 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019466 CAS Number: 873-94-9 EINECS: 212-855-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...