1-Tetracosanol ≥98.0%
Supplier: TCI America
T0593-5G
T0593-1G
TCT0593-005GEA
1199.22
CAD
TCT0593-005G
TCT0593-001G
1-Tetracosanol ≥98.0%
1-Tetracosanol
CAS Number: 506-51-4
MDL Number: MFCD00042662
Molecular Formula: C24H50O
Molecular Weight: 354.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 74
Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Lignoceryl Alcohol
Formula:
H₃C(CH₂)₂₂CH₂OH Boiling Pt: 190 °C (1013 hPa) Melting Pt: 74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042662 CAS Number: 506-51-4 EINECS: 208-043-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...