Tricosanoic acid ≥95.0%
Supplier: TCI America
|
Warning
|
T0402-1G
T0402-10G
TCT0402-001GEA
278.21
CAD
TCT0402-001G
TCT0402-010G
Tricosanoic acid ≥95.0%
Tricosanoic acid
CAS Number: 2433-96-7
MDL Number: MFCD00002808
Molecular Formula: C23H46O2
Molecular Weight: 354.62
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 80
Formula:
C₂₃H₄₆O₂ Boiling Pt: 399 °C (1013 hPa) Melting Pt: 80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002808 CAS Number: 2433-96-7 EINECS: 219-419-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...