1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0386-25G T0386-500G
TCT0386-025GEA 52.14 CAD
TCT0386-025G TCT0386-500G
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

CAS Number: 6001-64-5
MDL Number: MFCD00004461
Molecular Formula: C4H7Cl3O
Molecular Weight: 177.45
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 175
Melting point (°C): 78
Flash Point (°C): 100


Other Product Names / Synonyms: Acetonechloroform, beta,beta,beta-Trichloro-tert-butyl Alcohol, Chloretone

Formula: C₈H₁₆Cl₆O₃
Boiling Pt: 175 °C
Melting Pt: 78 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD02179352
CAS Number: 6001-64-5
EINECS: 200-317-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=899339f9f949b57f2cabce02d622667a8f97dce7&vid=899339f9f949b57f2cabce02d622667a8f97dce7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR