Tributyrin (glycerol tributyrate) ≥98.0%

Supplier: TCI America

T0364-500ML T0364-25ML
TCT0364-500MLEA 510.8 CAD
TCT0364-500ML TCT0364-025ML
Tributyrin (glycerol tributyrate) ≥98.0%
Tributyrin (Glycerol tributyrate)

CAS Number: 60-01-5
MDL Number: MFCD00009392
Molecular Formula: C15H26O6
Molecular Weight: 302.37
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Melting point (°C): -75
Flash Point (°C): 180
Specific Gravity (20/20): 1.04


Product Categories: Materials Chemistry, Resin Additives, Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tributyrate

Formula: C₁₅H₂₆O₆
Boiling Pt: 190 °C
Melting Pt: –75 °C
Density: 1.04 (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00009392
CAS Number: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f49c36d19f7dfde3734c1fee03c59fc28b6c4b9b&vid=f49c36d19f7dfde3734c1fee03c59fc28b6c4b9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR