Ethyl-p-toluate ≥99.0%
Supplier: TCI America
Synonyms:
Ethyl 4-methylbenzoate, Ethyl-4-methylbenzoate, 4-Methylbenzoic acid ethyl ester, 4-Toluic acid ethyl ester, p-Toluic acid ethyl ester
CAS Number: 94-08-6
MDL Number: MFCD00009117
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 232
Flash Point (°C): 118
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: p-Toluic Acid Ethyl Ester, 4-Methylbenzoic Acid Ethyl Ester, Ethyl 4-Methylbenzoate
Formula:
C₁₀H₁₂O₂ Boiling Pt: 232 °C Density: 1.03 (20 °C) Flash Pt: 118 °C |
MDL Number:
MFCD00009117 CAS Number: 94-08-6 EINECS: 202-301-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...